accessibility menu, dialog, popup

UserName

H2NNHNHNH2

CAS RN 4741-99-5

IUPAC Name: (2-aminoethyl)({3-[(2-aminoethyl)amino]propyl})amine
Synonyms: 1,4,8,11-tetraazaundecane 2,3,2-tetramine ethylenetrimethyleneethylenetetramine
Molecular Weight (g/mol): 160.27
Molecular Formula: C7H20N4
InChi Key: UWMHHZFHBCYGCV-UHFFFAOYSA-N
SMILES: NCCNCCCNCCN
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)