missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(2-aminoethyl)-1,3-propanediamine, 96%

Catalog No. AAH5495003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5495003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5495003 Supplier Thermo Scientific Chemicals Supplier No. H5495003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4741-99-5 | C7H20N4 | 160.265 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4741-99-5
Molecular Formula C7H20N4
Molecular Weight (g/mol) 160.265
MDL Number MFCD00008174
InChI Key UWMHHZFHBCYGCV-UHFFFAOYSA-N
Synonym n,n'-bis 2-aminoethyl-1,3-propanediamine, 1,4,8,11-tetraazaundecane, 2,3,2-tetramine, n,n'-bis 2-aminoethyl propane-1,3-diamine, 1,4,8,11-tetrazaundecane, ethylenetrimethyleneethylenetetramine, 1,3-propanediamine, n,n'-bis 2-aminoethyl, 1,9-diamino-3,7-diazanonane, n1,n1'-propane-1,3-diyl bis ethane-1,2-diamine, 3,7-diazanonane-1,9-diamine
PubChem CID 78479
ChEBI CHEBI:30348
IUPAC Name N,N'-bis(2-aminoethyl)propane-1,3-diamine
SMILES C(CNCCN)CNCCN

Specifications

Density 0.96
Boiling Point 142°C to 145°C
Flash Point 100°C (212°F)
Refractive Index 1.494
Linear Formula H2NCH2CH2NH(CH2)3NHCH2CH2NH2
Quantity 1 g
UN Number UN2735
Beilstein 1742732
Formula Weight 160.26
Percent Purity 97%
Chemical Name or Material N,N'-Bis(2-aminoethyl)-1,3-propanediamine
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: POLYAMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 225-254-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon; Store under Nitrogen

RUO – Research Use Only