missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(2-aminoethyl)-1,3-propanediamine 97.0+%, TCI America™

Catalog No. B14455G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B14455G Supplier TCI America Supplier No. B14455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4741-99-5
Molecular Formula C7H20N4
Molecular Weight (g/mol) 160.265
MDL Number MFCD00008174
InChI Key UWMHHZFHBCYGCV-UHFFFAOYSA-N
Synonym n,n'-bis 2-aminoethyl-1,3-propanediamine, 1,4,8,11-tetraazaundecane, 2,3,2-tetramine, n,n'-bis 2-aminoethyl propane-1,3-diamine, 1,4,8,11-tetrazaundecane, ethylenetrimethyleneethylenetetramine, 1,3-propanediamine, n,n'-bis 2-aminoethyl, 1,9-diamino-3,7-diazanonane, n1,n1'-propane-1,3-diyl bis ethane-1,2-diamine, 3,7-diazanonane-1,9-diamine
PubChem CID 78479
ChEBI CHEBI:30348
IUPAC Name N,N'-bis(2-aminoethyl)propane-1,3-diamine
SMILES C(CNCCN)CNCCN

Specifications

Color Yellow
Boiling Point 287°C
Quantity 5 g
UN Number 2735
Formula Weight 160.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N′-Bis(2-aminoethyl)-1,3-propanediamine
EINECSNumber (7)-0005
TSCA Yes