accessibility menu, dialog, popup

UserName

CAS RN 473923-97-6

IUPAC Name: 3-chloro-5-hydroxybenzonitrile
Synonyms: 3-chloro-5-hydroxybenzonitrile
Molecular Weight (g/mol): 153.57
Molecular Formula: C7H4ClNO
InChi Key: GHYUOOZZOMUNSY-UHFFFAOYSA-N
SMILES: OC1=CC(Cl)=CC(=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results