accessibility menu, dialog, popup

UserName

CAS RN 4701-17-1

IUPAC Name: 5-bromothiophene-2-carbaldehyde
Synonyms: 5-bromothiophene-2-carbaldehyde
Molecular Weight (g/mol): 191.04
Molecular Formula: C5H3BrOS
InChi Key: GFBVUFQNHLUCPX-UHFFFAOYSA-N
SMILES: BrC1=CC=C(S1)C=O
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)