accessibility menu, dialog, popup

UserName

CAS RN 4695-62-9

IUPAC Name: (1S,4R)-1,3,3-trimethylbicyclo[2.2.1]heptan-2-one
Synonyms: (+)-fenchone (+)-2-fenchanone (1S)-fenchone (1S,4R)-2-fenchanone d-fenchone trimethylbicyclo-2-heptanone (1S,4R)-fenchone (1S)-(+)-fenchone (+)-fenchan-2-one (1S,4R)-fenchan-2-one
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: LHXDLQBQYFFVNW-XCBNKYQSSA-N
SMILES: CC1(C)[C@@H]2CC[C@@](C)(C2)C1=O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)