accessibility menu, dialog, popup

UserName

CAS RN 461-17-6

IUPAC Name: 1,1,1-trifluoro-4-iodobutane
Synonyms: 1,1,1-trifluoro-4-iodobutane
Molecular Weight (g/mol): 237.99
Molecular Formula: C4H6F3I
InChi Key: LNDGACQEAYKNOI-UHFFFAOYSA-N
SMILES: FC(F)(F)CCCI
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)