accessibility menu, dialog, popup

UserName

FOHO

CAS RN 458-45-7

IUPAC Name: 3-(3-fluorophenyl)propanoic acid
Synonyms: 3-(3-fluorophenyl)propanoic acid
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: UBLMRADOKLXLCD-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC(F)=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)