missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Fluorophenyl)propionic acid, 97%

Catalog No. AAH2784706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2784706 5 g
AAH2784703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2784706 Supplier Thermo Scientific Chemicals Supplier No. H2784706
Only null left

CAS: 458-45-7 | C9H9FO2 | 168.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 458-45-7
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD01310821
InChI Key UBLMRADOKLXLCD-UHFFFAOYSA-N
PubChem CID 2737489
IUPAC Name 3-(3-fluorophenyl)propanoic acid
SMILES C1=CC(=CC(=C1)F)CCC(=O)O

Specifications

Melting Point 43°C to 47°C
Boiling Point 125°C (10 mmHg)
Flash Point 110°C (230°F)
Quantity 5 g
Synonym 3-3-fluorophenyl propionic acid, 3-3-fluorophenyl propanoic acid, 3-fluorohydrocinnamic acid, 3-3-fluoro-phenyl-propionic acid, 3-fluoro-benzenepropanoic acid, benzenepropanoic acid, 3-fluoro, pubchem13947, acmc-209k4f, 3-fluorobenzenepropanoic acid, benzenepropanoic acid,3-fluoro
Formula Weight 168.17
Percent Purity 97%
Chemical Name or Material 3-(3-Fluorophenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only