missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Fluorophenyl)propionic Acid 97.0+%, TCI America™

Catalog No. F07795G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07795G 5 g
F07791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07795G Supplier TCI America Supplier No. F07795G
Only null left

Chemical Identifiers

CAS 458-45-7
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD01310821
InChI Key UBLMRADOKLXLCD-UHFFFAOYSA-N
Synonym 3-3-fluorophenyl propionic acid, 3-3-fluorophenyl propanoic acid, 3-fluorohydrocinnamic acid, 3-3-fluoro-phenyl-propionic acid, 3-fluoro-benzenepropanoic acid, benzenepropanoic acid, 3-fluoro, pubchem13947, acmc-209k4f, 3-fluorobenzenepropanoic acid, benzenepropanoic acid,3-fluoro
PubChem CID 2737489
IUPAC Name 3-(3-fluorophenyl)propanoic acid
SMILES C1=CC(=CC(=C1)F)CCC(=O)O

Specifications

Melting Point 43°C
Boiling Point 119°C
Quantity 5 g
Formula Weight 168.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(3-Fluorophenyl)propionic Acid