accessibility menu, dialog, popup

UserName

SNH

CAS RN 4556-23-4

IUPAC Name: 1,4-dihydropyridine-4-thione
Synonyms: 4(1H)-pyridinethione
Molecular Weight (g/mol): 111.16
Molecular Formula: C5H5NS
InChi Key: FHTDDANQIMVWKZ-UHFFFAOYSA-N
SMILES: S=C1C=CNC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)