missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptopyridine 97.0+%, TCI America™

Catalog No. M06305G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M06305G 5 g
M063025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M06305G Supplier TCI America Supplier No. M06305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4556-23-4 | C5H5NS | 111.16 g/mol

Chemical Identifiers

CAS 4556-23-4
Molecular Formula C5H5NS
Molecular Weight (g/mol) 111.16
MDL Number MFCD00006422
InChI Key FHTDDANQIMVWKZ-UHFFFAOYSA-N
Synonym 4-mercaptopyridine, pyridine-4-thiol, 4-pyridinethiol, 4-thiopyridine, 4-pyridinethione, pyridine, 4-mercapto, 4-thiopyridone, 4 1h-pyridinethione, pyridine-4 1h-thione, 5-21-07-00167 beilstein handbook reference
PubChem CID 2723889
IUPAC Name 1,4-dihydropyridine-4-thione
SMILES S=C1C=CNC=C1

Specifications

Melting Point 180°C
Quantity 5 g
Formula Weight 111.16
Percent Purity ≥97.0% (T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 4-Mercaptopyridine
RTECSNumber UT8578000
TSCA No
Recommended Storage Freezer