accessibility menu, dialog, popup

UserName

NH2FOHO

CAS RN 455-87-8

IUPAC Name: 4-amino-3-fluorobenzoic acid
Synonyms: 4-amino-3-fluorobenzoic acid
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: JSKXHTHMCCDEGD-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1F)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)