CAS RN 4549-31-9
CAS RN 4549-31-9
IUPAC Name:
1,7-dibromoheptane
Synonyms:
heptane, 1,7-dibromo-
Molecular Weight (g/mol):
258
Molecular Formula:
C7H14Br2
InChi Key:
LVWSZGCVEZRFBT-UHFFFAOYSA-N
SMILES:
BrCCCCCCCBr
1
–
1
of
1
results
1
1,7-Dibromoheptane 98.0+%, TCI America™
CAS: 4549-31-9 Molecular Formula: C7H14Br2 Molecular Weight (g/mol): 258.00 MDL Number: MFCD00000274 InChI Key: LVWSZGCVEZRFBT-UHFFFAOYSA-N Synonym: heptane, 1,7-dibromo,heptamethylene dibromide,heptane,7-dibromo,1,7-dibromo-heptane,pubchem3887,dibromo-1,7 heptane,acmc-209k2q,1,7-dibromoheptane,ksc236m7h,lvwszgcvezrfbt-uhfffaoysa PubChem CID: 78309 IUPAC Name: 1,7-dibromoheptane SMILES: BrCCCCCCCBr