accessibility menu, dialog, popup

UserName

CAS RN 4536-23-6

IUPAC Name: (2R)-2-methylhexanoate
Synonyms: (2R)-2-methylhexanoate
Molecular Weight (g/mol): 129.18
Molecular Formula: C7H13O2
InChi Key: CVKMFSAVYPAZTQ-ZCFIWIBFSA-M
SMILES: CCCC[C@@H](C)C([O-])=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)