missing translation for 'onlineSavingsMsg'
Learn More

2-Methylhexanoic Acid 98.0+%, TCI America™

Catalog No. M09475ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09475ML 5 mL
M094725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09475ML Supplier TCI America Supplier No. M09475ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4536-23-6
Molecular Formula C7H13O2
Molecular Weight (g/mol) 129.18
MDL Number MFCD00002674
InChI Key CVKMFSAVYPAZTQ-ZCFIWIBFSA-M
Synonym 2-methylcaproic acid, hexanoic acid, 2-methyl, methylhexanoic acid, 2-methyl hexanoic acid, 2-hexanecarboxylic acid, alpha-methylcaproic acid, 2-methyl-hexanoic acid, fema no. 3191, .alpha.-methylcaproic acid, hexanoic acid, methyl
PubChem CID 20653
IUPAC Name (2R)-2-methylhexanoate
SMILES CCCC[C@@H](C)C([O-])=O

Specifications

Color Yellow
Boiling Point 72°C
Quantity 5 mL
UN Number 3265
Formula Weight 130.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methylhexanoic Acid
EINECSNumber (2)-0608
TSCA Yes