missing translation for 'onlineSavingsMsg'
Learn More

2-Methylhexanoic acid, 98%

Catalog No. AAA1129014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1129014 25 g
AAA1129006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1129014 Supplier Thermo Scientific Chemicals Supplier No. A1129014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4536-23-6 | C7H13O2 | 129.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4536-23-6
Molecular Formula C7H13O2
Molecular Weight (g/mol) 129.18
MDL Number MFCD00002674
InChI Key CVKMFSAVYPAZTQ-ZCFIWIBFSA-M
Synonym 2-methylcaproic acid, hexanoic acid, 2-methyl, methylhexanoic acid, 2-methyl hexanoic acid, 2-hexanecarboxylic acid, alpha-methylcaproic acid, 2-methyl-hexanoic acid, fema no. 3191, .alpha.-methylcaproic acid, hexanoic acid, methyl
PubChem CID 20653
SMILES CCCC[C@@H](C)C([O-])=O

Specifications

Density 0.916
Boiling Point 209°C to 210°C
Flash Point 105°C (221°F)
Odor Rancid
Refractive Index 1.4205
Quantity 25 g
UN Number UN3265
Beilstein 1721227
IUPAC Name (2R)-2-methylhexanoate
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material 2-Methylhexanoic acid
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 224-883-9
RTECSNumber MO8400600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only