missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ 2-Chloro-4-Fluorotoluene, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111022780
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022780 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022780 Supplier Merck Emd Millipore Supplier No. 80730500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

Specifications

CAS 452-73-3
Density 1.197 g/mL (at 25°C (literature))
Boiling Point 154°C to 156°C (literature)
Flash Point 50°C (Closed Cup)
Molecular Formula C7H6ClF
Linear Formula CH3C6H3(Cl)F
Refractive Index n20/D 1.499 (literature)
Quantity 500 mg
MDL Number MFCD00000572
UN Number UN 1993 C 3 / PGIII
Molecular Weight (g/mol) 144.58
Formula Weight 144.57
Grade Trace CERT
Physical Form Liquid
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC