accessibility menu, dialog, popup

UserName

CAS RN 452-66-4

CH3ClF

CAS RN 452-66-4

IUPAC Name: 4-chloro-1-fluoro-2-methylbenzene
Synonyms: 4-chloro-1-fluoro-2-methylbenzene
Molecular Weight (g/mol): 144.57
Molecular Formula: C7H6ClF
InChi Key: JOXXHDGUTVUBDL-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results