missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluorotoluene 97.0+%, TCI America™

Catalog No. C15875G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C15875G Supplier TCI America Supplier No. C15875G
Only null left

Chemical Identifiers

CAS 452-66-4
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.573
MDL Number MFCD00018504
InChI Key JOXXHDGUTVUBDL-UHFFFAOYSA-N
Synonym 5-chloro-2-fluorotoluene, 2-fluoro-5-chlorotoluene, benzene, 4-chloro-1-fluoro-2-methyl, pubchem1609, acmc-209k1f, 5-chloro-2-fluoro-toluene, ksc493m8d, joxxhdgutvubdl-uhfffaoysa, 1-chloro-4-fluoro-3-methylbenzene
PubChem CID 2725113
IUPAC Name 4-chloro-1-fluoro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Cl)F

Specifications

Color Colorless
Quantity 5 g
UN Number 1993
Formula Weight 144.57
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Chloro-2-fluorotoluene