accessibility menu, dialog, popup

UserName

CH3HOOH

CAS RN 4457-71-0

IUPAC Name: 3-methylpentane-1,5-diol
Synonyms: 1,5-pentanediol, 3-methyl-
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: SXFJDZNJHVPHPH-UHFFFAOYSA-N
SMILES: CC(CCO)CCO
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)