missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1,5-pentanediol 97.0+%, TCI America™

Catalog No. M0710500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0710500ML 500 mL
M071025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0710500ML Supplier TCI America Supplier No. M0710500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4457-71-0
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00039547
InChI Key SXFJDZNJHVPHPH-UHFFFAOYSA-N
PubChem CID 20524
IUPAC Name 3-methylpentane-1,5-diol
SMILES CC(CCO)CCO

Specifications

Color Colorless
Boiling Point 150°C
Quantity 500 mL
Synonym 3-methyl-1,5-pentanediol, 1,5-pentanediol, 3-methyl, 1,5-dihydroxy-3-methylpentane, unii-88495m4hns, acmc-1ai92, 3-methyl-1,5-pentane diol, 3-methyl-pentane-1,5-diol, 3-methylpentane-1,5-diol, 1,5-pentanediol,3-methyl, 4-01-00-02571 beilstein handbook reference
Formula Weight 118.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-1,5-pentanediol
EINECSNumber (2)-0240
RTECSNumber SA0800000
TSCA Yes