missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1,5-pentanediol, 99%

Catalog No. AAB2054614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2054614 25 g
AAB2054606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2054614 Supplier Thermo Scientific Chemicals Supplier No. B2054614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4457-71-0 | C6H14O2 | 118.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4457-71-0
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00039547
InChI Key SXFJDZNJHVPHPH-UHFFFAOYSA-N
Synonym 3-methyl-1,5-pentanediol, 1,5-pentanediol, 3-methyl, 1,5-dihydroxy-3-methylpentane, unii-88495m4hns, acmc-1ai92, 3-methyl-1,5-pentane diol, 3-methyl-pentane-1,5-diol, 3-methylpentane-1,5-diol, 1,5-pentanediol,3-methyl, 4-01-00-02571 beilstein handbook reference
PubChem CID 20524
IUPAC Name 3-methylpentane-1,5-diol
SMILES CC(CCO)CCO

Specifications

Density 0.97
Flash Point 143°C (289°F)
Refractive Index 1.454
Quantity 25 g
Beilstein 1697331
Formula Weight 118.18
Percent Purity 99%
Chemical Name or Material 3-Methyl-1,5-pentanediol
EINECSNumber 224-709-1
RTECSNumber SA0800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only