accessibility menu, dialog, popup

UserName

HOHO

CAS RN 4370-02-9

IUPAC Name: 2,3-dihydro-1H-indene-1,2-diol
Synonyms: 2,3-dihydro-1H-indene-1,2-diol
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: YKXXBEOXRPZVCC-UHFFFAOYNA-N
SMILES: OC1CC2=CC=CC=C2C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results