accessibility menu, dialog, popup

UserName

CAS RN 426844-51-1

IUPAC Name: 4,4-difluoropyrrolidine-2-carboxamide hydrochloride
Synonyms: 4,4-difluoropyrrolidine-2-carboxamide hydrochloride
Molecular Weight (g/mol): 186.59
Molecular Formula: C5H9ClF2N2O
InChi Key: RRQDQYAEZGKHOB-UHFFFAOYNA-N
SMILES: Cl.NC(=O)C1CC(F)(F)CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results