missing translation for 'onlineSavingsMsg'
Learn More

4,4-Difluoro-L-prolinamide hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. AAH2777003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2777003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2777003 Supplier Thermo Scientific Chemicals Supplier No. H2777003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 426844-51-1
Molecular Formula C5H9ClF2N2O
Molecular Weight (g/mol) 186.59
MDL Number MFCD06797014
InChI Key RRQDQYAEZGKHOB-UHFFFAOYNA-N
Synonym 4,4-difluoro-l-prolinamide hydrochloride, s-4,4-difluoropyrrolidine-2-carboxamide hydrochloride, s-4,4-difluoropyrolidine-2-carboxamide hydrochloride, 2s-4,4-difluoropyrrolidine-2-carboxamide hydrochloride, c5h8f2n2o.hcl, 4,4-difluoro-l-prolinamide hcl, 4,4-difluoro-l-prolinamide, hcl, 4,4-difluoro-l-prolinamide-hydrogen chloride 1/1, 2s-4,4-difluoro-2-pyrrolidinecarboxamide hydrochloride
PubChem CID 45158909
IUPAC Name (2S)-4,4-difluoropyrrolidine-2-carboxamide;hydrochloride
SMILES Cl.NC(=O)C1CC(F)(F)CN1

Specifications

Melting Point 221°C to 223°C (decomposition)
Quantity 1 g
Optical Rotation −31° (c=1 in Water)
Formula Weight 186.59
Percent Purity 98%
Chemical Name or Material 4,4-Difluoro-L-prolinamide hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only