accessibility menu, dialog, popup

UserName

SOOFFF

CAS RN 426-58-4

IUPAC Name: trifluoromethanesulfonylbenzene
Synonyms: sulfone,phenyl-trifluoromethyl
Molecular Weight (g/mol): 210.17
Molecular Formula: C7H5F3O2S
InChi Key: UPGBQYFXKAKWQC-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results