accessibility menu, dialog, popup

UserName

HO

CAS RN 4254-29-9

IUPAC Name: 2,3-dihydro-1H-inden-2-ol
Synonyms: 2,3-dihydro-1H-inden-2-ol
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: KMGCKSAIIHOKCX-UHFFFAOYSA-N
SMILES: OC1CC2=CC=CC=C2C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results