CAS RN 424-40-8
CAS RN 424-40-8
IUPAC Name:
1,5-diethyl 2,2,3,3,4,4-hexafluoropentanedioate
Synonyms:
1,5-diethyl 2,2,3,3,4,4-hexafluoropentanedioate
Molecular Weight (g/mol):
296.17
Molecular Formula:
C9H10F6O4
InChi Key:
MSDPXVBLFJODJO-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(=O)OCC
1
–
1
of
1
results
1
Diethyl 2,2,3,3,4,4-Hexafluoropentanedioate 98.0+%, TCI America™
CAS: 424-40-8 Molecular Formula: C9H10F6O4 Molecular Weight (g/mol): 296.17 MDL Number: MFCD00015156 InChI Key: MSDPXVBLFJODJO-UHFFFAOYSA-N Synonym: diethyl hexafluoroglutarate,diethyl perfluoroglutarate,diethyl hexafluoropentanedioate,hexafluoroglutaric acid diethyl ester,pentanedioic acid, hexafluoro-, diethyl ester,glutaric acid, hexafluoro-, diethyl ester,hexafluoropentanedioic acid diethyl ester,diethylester kyseliny hexafluorglutarove,diethylester kyseliny hexafluorglutarove czech,pentanedioic acid, 2,2,3,3,4,4-hexafluoro-, 1,5-diethyl ester PubChem CID: 9874 IUPAC Name: 1,5-diethyl 2,2,3,3,4,4-hexafluoropentanedioate SMILES: CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(=O)OCC