missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2,2,3,3,4,4-Hexafluoropentanedioate 98.0+%, TCI America™

Catalog No. H07441G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H07441G Supplier TCI America Supplier No. H07441G
Only null left

Chemical Identifiers

CAS 424-40-8
Molecular Formula C9H10F6O4
Molecular Weight (g/mol) 296.17
MDL Number MFCD00015156
InChI Key MSDPXVBLFJODJO-UHFFFAOYSA-N
Synonym diethyl hexafluoroglutarate, diethyl perfluoroglutarate, diethyl hexafluoropentanedioate, hexafluoroglutaric acid diethyl ester, pentanedioic acid, hexafluoro-, diethyl ester, glutaric acid, hexafluoro-, diethyl ester, hexafluoropentanedioic acid diethyl ester, diethylester kyseliny hexafluorglutarove, diethylester kyseliny hexafluorglutarove czech, pentanedioic acid, 2,2,3,3,4,4-hexafluoro-, 1,5-diethyl ester
PubChem CID 9874
IUPAC Name 1,5-diethyl 2,2,3,3,4,4-hexafluoropentanedioate
SMILES CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(=O)OCC

Specifications

Color Yellow
Boiling Point 84°C
Quantity 1 g
Formula Weight 296.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 2,2,3,3,4,4-Hexafluoropentanedioate
RTECSNumber MA3750500
TSCA Yes