CAS RN 42333-76-6
IUPAC Name:
tris(pyridin-3-yl)-1,3,5-triazine
Synonyms:
tris(pyridin-3-yl)-1,3,5-triazine
Molecular Weight (g/mol):
312.34
Molecular Formula:
C18H12N6
InChi Key:
UOSYDSDIRAXTTF-UHFFFAOYSA-N
SMILES:
C1=CC(=CN=C1)C1=NC(=NC(=N1)C1=CN=CC=C1)C1=CN=CC=C1
1
–
1
of
1
results
1
Promotions Available
2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%
2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%
CAS: 42333-76-6 Molecular Formula: C18H12N6 Molecular Weight (g/mol): 312.34 MDL Number: MFCD23379843 InChI Key: UOSYDSDIRAXTTF-UHFFFAOYSA-N Synonym: 2,4,6-tris 3-pyridyl-s-triazine,2,4,6-tri-3-pyridinyl-1,3,5-triazine,tris pyridin-3-yl-1,3,5-triazine,2,4,6-tris 3-pyridyl-1,3,5-triazine PubChem CID: 11472427 IUPAC Name: 2,4,6-tripyridin-3-yl-1,3,5-triazine SMILES: C1=CC(=CN=C1)C1=NC(=NC(=N1)C1=CN=CC=C1)C1=CN=CC=C1