accessibility menu, dialog, popup

UserName

CAS RN 4233-96-9

IUPAC Name: (2S,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3-yl 3,4,5-trihydroxybenzoate
Synonyms: gallocatechin gallate
Molecular Weight (g/mol): 458.38
Molecular Formula: C22H18O11
InChi Key: WMBWREPUVVBILR-NQIIRXRSSA-N
SMILES: OC1=CC(O)=C2C[C@@H](OC(=O)C3=CC(O)=C(O)C(O)=C3)[C@@H](OC2=C1)C1=CC(O)=C(O)C(O)=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results