missing translation for 'onlineSavingsMsg'
Learn More

(-)-Gallocatechin gallate

Catalog No. AAJ62740MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62740MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62740MA Supplier Thermo Scientific Chemicals Supplier No. J62740MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4233-96-9 | C22H18O11 | 458.375 g/mol

An antioxidant from green tea

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4233-96-9
Molecular Formula C22H18O11
Molecular Weight (g/mol) 458.375
MDL Number MFCD00214298
InChI Key WMBWREPUVVBILR-NQIIRXRSSA-N
Synonym =--gallocatechin gallate, gallocatechin gallate, =--gallocatechol gallate, unii-irw3c4y31q, gallocatechin gallate gcg, irw3c4y31q, gallocatechin gallate,-, =--gallocatechin-3-o-gallate, 2s,3r-5,7-dihydroxy-2-3,4,5-trihydroxyphenyl chroman-3-yl 3,4,5-trihydroxybenzoate, 2s,3r-2-3,4,5-trihydroxyphenyl-3,4-dihydro-1 2h-benzopyran-3,5,7-triol 3-3,4,5-trihydroxybenzoate
PubChem CID 199472
IUPAC Name [(2S,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O

Specifications

Color White
Quantity 10 mg
Formula Weight 458.38
Physical Form Powder
Chemical Name or Material (-)-Gallocatechin gallate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber DH9000000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only