accessibility menu, dialog, popup

UserName

CAS RN 41757-95-3

IUPAC Name: 1-(2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecin-15-yl)ethan-1-one
Synonyms: 1-(2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecin-15-yl)ethanone
Molecular Weight (g/mol): 310.35
Molecular Formula: C16H22O6
InChi Key: XNJYHEGDUAQGEE-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2OCCOCCOCCOCCOC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results