missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Ciclopirox Olamine USP
Description
Ciclopirox Olamine USP is a white to slightly yellowish-white, crystalline powder.
Specifications
Specifications
| Chemical Name or Material | Ciclopirox Olamine USP |
| Packaging | Jar 300ml, White, HDPE, Round, 53-400, TRIVENI-DB, CT |
| CAS | 41621-49-2 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 268.3546 |
| Quantity | 25 g |
| Percent Purity | 97.5 to 101.5 % |
| Grade | USP |