accessibility menu, dialog, popup

UserName

CAS RN 41484-35-9

H3CH3CCH3OOSOOOHCH3H3CCH3CH3CH3H3CHOCH3H3CCH3

CAS RN 41484-35-9

IUPAC Name: 2-[(2-{[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]oxy}ethyl)sulfanyl]ethyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate
Synonyms: 2-[(2-{[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]oxy}ethyl)sulfanyl]ethyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate
Molecular Weight (g/mol): 642.94
Molecular Formula: C38H58O6S
InChi Key: VFBJXXJYHWLXRM-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(CCC(=O)OCCSCCOC(=O)CCC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results