accessibility menu, dialog, popup

UserName

CAS RN 4138-26-5

H2NO(R)H2N

CAS RN 4138-26-5

IUPAC Name: (3R)-3-carbamoylpiperidin-1-ium
Synonyms: (R)-nipecotamide(1+) (3R)-piperidine-3-carboxamide (R)-nipecotamide cation (3R)-hexahydronicotinamide(1+)
Molecular Weight (g/mol): 129.18
Molecular Formula: C6H13N2O
InChi Key: BVOCPVIXARZNQN-RXMQYKEDSA-O
SMILES: NC(=O)[C@@H]1CCC[NH2+]C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)