missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nipecotamide 98.0+%, TCI America™

$64.38 - $171.14

Chemical Identifiers

CAS 4138-26-5
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.175
MDL Number MFCD00005993
InChI Key BVOCPVIXARZNQN-UHFFFAOYSA-N
Synonym 3-piperidine carboxamide, 3-piperidinecarboxamide, acmc-20lak2, hexahydronicotinamide, nipecotamide, nipecotic acid amide, piperidine-3-carboxylic acid amide, pubchem11797, racemic nipecotamide
PubChem CID 92980
ChEBI CHEBI:60116
IUPAC Name piperidine-3-carboxamide
SMILES C1CC(CNC1)C(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N00975G View Documents Tci America
N00975G
5g Each for $64.38
 
N009725G View Documents Tci America
N009725G
25g Each for $171.14
 
Specifications

Chemical Identifiers

4138-26-5
128.175
BVOCPVIXARZNQN-UHFFFAOYSA-N
92980
piperidine-3-carboxamide
C6H12N2O
MFCD00005993
3-piperidine carboxamide, 3-piperidinecarboxamide, acmc-20lak2, hexahydronicotinamide, nipecotamide, nipecotic acid amide, piperidine-3-carboxylic acid amide, pubchem11797, racemic nipecotamide
CHEBI:60116
C1CC(CNC1)C(=O)N

Specifications

Undesignated
5g
C6H12N2O
3-piperidine carboxamide, 3-piperidinecarboxamide, acmc-20lak2, hexahydronicotinamide, nipecotamide, nipecotic acid amide, piperidine-3-carboxylic acid amide, pubchem11797, racemic nipecotamide
C1CC(CNC1)C(=O)N
128.175
CHEBI:60116
Crystal-Powder at 20°C
Nipecotamide
111°C
4138-26-5
MFCD00005993
BVOCPVIXARZNQN-UHFFFAOYSA-N
piperidine-3-carboxamide
92980
128.18
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No