missing translation for 'onlineSavingsMsg'
Learn More

Nipecotamide, 95%, Thermo Scientific™

Chemical Identifiers

CAS 4138-26-5
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.17
MDL Number MFCD00005993
InChI Key BVOCPVIXARZNQN-UHFFFAOYSA-N
Synonym nipecotamide, 3-piperidinecarboxamide, nipecotic acid amide, hexahydronicotinamide, piperidine-3-carboxylic acid amide, racemic nipecotamide, pubchem11797, acmc-20lak2, 3-piperidine carboxamide
PubChem CID 92980
ChEBI CHEBI:60116
IUPAC Name piperidine-3-carboxamide
SMILES C1CC(CNC1)C(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC128300050
SDS View Documents
Thermo Scientific Chemicals
128300050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC128300500
SDS View Documents
Thermo Scientific Chemicals
128300500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4138-26-5
128.17
BVOCPVIXARZNQN-UHFFFAOYSA-N
92980
piperidine-3-carboxamide
C6H12N2O
MFCD00005993
nipecotamide, 3-piperidinecarboxamide, nipecotic acid amide, hexahydronicotinamide, piperidine-3-carboxylic acid amide, racemic nipecotamide, pubchem11797, acmc-20lak2, 3-piperidine carboxamide
CHEBI:60116
C1CC(CNC1)C(=O)N

Specifications

4138-26-5
100.0
95%
C6H12N2O
5g
BVOCPVIXARZNQN-UHFFFAOYSA-N
piperidine-3-carboxamide
92980
128.17
Nipecotamide, 95%
94.0
Authentic
Glass bottle
MFCD00005993
nipecotamide, 3-piperidinecarboxamide, nipecotic acid amide, hexahydronicotinamide, piperidine-3-carboxylic acid amide, racemic nipecotamide, pubchem11797, acmc-20lak2, 3-piperidine carboxamide
C1CC(CNC1)C(=O)N
128.17
CHEBI:60116
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

EINECSNumber : 223-962-5

SDS
Documents

Documents

RUO – Research Use Only