accessibility menu, dialog, popup

UserName

CAS RN 4130-54-5

IUPAC Name: octadecanehydrazide
Synonyms: octadecanehydrazide
Molecular Weight (g/mol): 298.52
Molecular Formula: C18H38N2O
InChi Key: BYTFESSQUGDMQQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)NN
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)