missing translation for 'onlineSavingsMsg'
Learn More

Stearic Hydrazide 95.0+%, TCI America™

Catalog No. S022225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S022225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S022225G Supplier TCI America Supplier No. S022225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4130-54-5
Molecular Formula C18H38N2O
Molecular Weight (g/mol) 298.515
MDL Number MFCD00014764
InChI Key BYTFESSQUGDMQQ-UHFFFAOYSA-N
Synonym stearic acid hydrazide, stearic hydrazide, stearohydrazide, octadecanoic acid, hydrazide, stearoylhydrazine, stearic acid, hydrazide, unii-3fv1c68xop, 3fv1c68xop, octadecanohydrazide #, stearic acid hydrazone
PubChem CID 20088
IUPAC Name octadecanehydrazide
SMILES CCCCCCCCCCCCCCCCCC(=O)NN

Specifications

Melting Point 116°C
Color White
Quantity 25 g
Formula Weight 298.52
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Stearic Hydrazide