accessibility menu, dialog, popup

UserName

H3CNHNHH3CHNCH3NHCH3

CAS RN 41203-22-9

IUPAC Name: 1,4,8,11-tetramethyl-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraium
Synonyms: 1,4,8,11-tetramethyl-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraium
Molecular Weight (g/mol): 260.47
Molecular Formula: C14H36N4
InChi Key: HRFJEOWVAGSJNW-UHFFFAOYNA-R
SMILES: C[NH+]1CCC[NH+](C)CC[NH+](C)CCC[NH+](C)CC1
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)