missing translation for 'onlineSavingsMsg'
Learn More

1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane 98.0+%, TCI America™

Catalog No. T1877200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1877200MG 200 mg
T18771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1877200MG Supplier TCI America Supplier No. T1877200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41203-22-9
Molecular Formula C14H36N4
Molecular Weight (g/mol) 260.47
MDL Number MFCD00005106
InChI Key HRFJEOWVAGSJNW-UHFFFAOYNA-R
PubChem CID 123502
IUPAC Name 1,4,8,11-tetramethyl-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraium
SMILES C[NH+]1CCC[NH+](C)CC[NH+](C)CCC[NH+](C)CC1

Specifications

Melting Point 40°C
Color White-Yellow
Quantity 200 mg
Synonym 1,4,8,11-tetramethyl-1,4,8,11-tetraazacyclotetradecane, tetramethylcyclam, 1,4,8,11-tetraazacyclotetradecane, 1,4,8,11-tetramethyl, acmc-1ankx, 1,4,8,11-tetramethylcyclam, 1,4,8,11-tetraazacyclotetradecane,1,4,8,11-tetramethyl, 1,4,8,11-tetramethyl-1,4,8, 11-tetraazacyclotetradecane, 1r,4s,8r,11s-1,4,8,11-tetramethyl-1,4,8,11-tetraazacyclotetradecane, 1alpha,4beta,8beta,11alpha-tetramethyl-1,4,8,11-tetraazacyclotetradecane
Formula Weight 256.44
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane
RTECSNumber XA5255700
TSCA No