missing translation for 'onlineSavingsMsg'
Learn More

1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane

Catalog No. AA3085403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3085403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3085403 Supplier Thermo Scientific Chemicals Supplier No. 03085403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 41203-22-9 | C14H36N4 | 260.47 g/mol

1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane is used as a macrocyclic chelating agent in coordination chemistry. It is also used as a ligand which bind strongly to a wide range of metal ions and as crown ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41203-22-9
Molecular Formula C14H36N4
Molecular Weight (g/mol) 260.47
MDL Number MFCD00005106
InChI Key HRFJEOWVAGSJNW-UHFFFAOYNA-R
Synonym 1,4,8,11-tetramethyl-1,4,8,11-tetraazacyclotetradecane, tetramethylcyclam, 1,4,8,11-tetraazacyclotetradecane, 1,4,8,11-tetramethyl, acmc-1ankx, 1,4,8,11-tetramethylcyclam, 1,4,8,11-tetraazacyclotetradecane,1,4,8,11-tetramethyl, 1,4,8,11-tetramethyl-1,4,8, 11-tetraazacyclotetradecane, 1r,4s,8r,11s-1,4,8,11-tetramethyl-1,4,8,11-tetraazacyclotetradecane, 1alpha,4beta,8beta,11alpha-tetramethyl-1,4,8,11-tetraazacyclotetradecane
PubChem CID 123502
IUPAC Name 1,4,8,11-tetramethyl-1,4,8,11-tetrazacyclotetradecane
SMILES C[NH+]1CCC[NH+](C)CC[NH+](C)CCC[NH+](C)CC1

Specifications

Melting Point 34°C to 36°C
Flash Point >110°C (230°F)
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 256.43
Physical Form Crystalline
Chemical Name or Material 1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only