missing translation for 'onlineSavingsMsg'
Learn More

2-Decyn-1-ol, 97%

Catalog No. AAL0552314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0552314 25 g
AAL0552306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0552314 Supplier Thermo Scientific Chemicals Supplier No. L0552314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4117-14-0 | C10H18O | 154.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4117-14-0
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00041567
InChI Key GFLHGTKUCYPXHB-UHFFFAOYSA-N
Synonym 2-decyn-1-ol, 2-decyne-1-ol, n-heptylhydroxymethylacetylene, unii-l4949b9xh9, decynol, pubchem11035, acmc-1arrx, heptyl hydroxymethyl acetylene, 2-decyn-1-ol,
PubChem CID 77763
IUPAC Name dec-2-yn-1-ol
SMILES CCCCCCCC#CCO

Specifications

Density 0.855
Boiling Point 114°C to 116°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4585
Quantity 25 g
Beilstein 1852842
Formula Weight 154.25
Percent Purity 97%
Chemical Name or Material 2-Decyn-1-ol
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only