CAS RN 4093-31-6
IUPAC Name:
methyl 5-chloro-4-acetamido-2-methoxybenzoate
Synonyms:
methyl 5-chloro-4-acetamido-2-methoxybenzoate
Molecular Weight (g/mol):
257.67
Molecular Formula:
C11H12ClNO4
InChi Key:
OUEXNQRVYGYGIK-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(Cl)=C(NC(C)=O)C=C1OC
1
–
1
of
1
results
1
Metoclopramide Related Compound B, Certified Reference Material, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.