missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metoclopramide Related Compound B, Certified Reference Material, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 4093-31-6 |
| Molecular Formula | C11H12ClNO4 |
| Linear Formula | CH3CONHC6H2(Cl)(OCH3)CO2CH3 |
| MDL Number | MFCD00048191 |
| Melting Point | 153°C to 156°C (literature) |
| Synonym | Methyl 4-acetamido-5-chloro-2-methoxybenzoate |
| Quantity | 100 mg |
| Formula Weight | 257.67 |
| Grade | Certified Reference Material |