accessibility menu, dialog, popup

UserName

CH3OOClH2N

CAS RN 40872-87-5

IUPAC Name: methyl 3-amino-4-chlorobenzoate
Synonyms: methyl 3-amino-4-chlorobenzoate
Molecular Weight (g/mol): 185.61
Molecular Formula: C8H8ClNO2
InChi Key: LOCJPOYKBUUVKU-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(Cl)C(N)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results