accessibility menu, dialog, popup

UserName

NH2HOO

CAS RN 40851-65-8

IUPAC Name: 2-[2-(aminomethyl)phenyl]acetic acid
Synonyms: [2-(aminomethyl)phenyl]acetic acid
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: VLOIVYPDUSVCLZ-UHFFFAOYSA-N
SMILES: NCC1=CC=CC=C1CC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results