accessibility menu, dialog, popup

UserName

CH3OCH3OH3CO

CAS RN 4077-47-8

IUPAC Name: 4-methoxy-2,5-dimethyl-2,3-dihydrofuran-3-one
Synonyms: 4-methoxy-2,5-dimethyl-2H-furan-3-one
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: SIMKGHMLPVDSJE-UHFFFAOYNA-N
SMILES: COC1=C(C)OC(C)C1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results